Assay Details
| Gene Symbol: LOC101108113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963224.1 | XP_027819025.1 | 31 - 32 | 3406 | 87 | |
| XM_027963226.1 | XP_027819027.1 | 31 - 32 | 3412 | 87 | ||
More Information
Set Membership: |
|

LOC101108113
Genomic Map