Assay Details
| Gene Symbol: LOC101108113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963224.1 | XP_027819025.1 | 9 - 10 | 1031 | 72 | |
| XM_027963225.1 | XP_027819026.1 | 9 - 10 | 1031 | 72 | ||
| XM_027963226.1 | XP_027819027.1 | 9 - 10 | 1037 | 72 | ||
More Information
Set Membership: |
|

LOC101108113
Genomic Map