Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006608.3 | XP_004006657.1 | 2 - 3 | 328 | 76 | |
| XM_012174788.3 | XP_012030178.1 | 2 - 3 | 256 | 76 | ||
More Information
Set Membership: |
|

ORMDL2
Genomic Map