Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012188639.3 | XP_012044029.2 | 3 - 4 | 1057 | 90 | |
| XM_027976805.1 | XP_027832606.1 | 3 - 4 | 781 | 90 | ||
More Information
Set Membership: |
|

KLF6
Genomic Map