Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013035.3 | XP_004013084.1 | 6 - 7 | 793 | 66 | |
| XM_012186345.2 | XP_012041735.1 | 6 - 7 | 821 | 66 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map