Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980079.1 | XP_027835880.1 | 5 - 6 | 1028 | 79 | |
| XM_027980080.1 | XP_027835881.1 | 5 - 6 | 858 | 79 | ||
| XM_027980081.1 | XP_027835882.1 | 5 - 6 | 722 | 79 | ||
More Information
Set Membership: |
|

PAIP1
Genomic Map