Assay Details
| Gene Symbol: RAB11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008567.2 | XP_004008616.1 | 4 - 5 | 725 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB11B| Gene Symbol: RAB11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008567.2 | XP_004008616.1 | 4 - 5 | 725 | 87 | |