Assay Details
| Gene Symbol: LOC101120580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971620.1 | XP_027827421.1 | 17 - 18 | 2178 | 70 | |
| XM_027971621.1 | XP_027827422.1 | 18 - 19 | 2444 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120580| Gene Symbol: LOC101120580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971620.1 | XP_027827421.1 | 17 - 18 | 2178 | 70 | |
| XM_027971621.1 | XP_027827422.1 | 18 - 19 | 2444 | 70 | ||