Assay Details
| Gene Symbol: LOC101120580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971620.1 | XP_027827421.1 | 7 - 8 | 648 | 88 | |
| XM_027971621.1 | XP_027827422.1 | 8 - 9 | 914 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120580| Gene Symbol: LOC101120580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971620.1 | XP_027827421.1 | 7 - 8 | 648 | 88 | |
| XM_027971621.1 | XP_027827422.1 | 8 - 9 | 914 | 88 | ||