Assay Details
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187003.3 | XP_012042393.1 | 8 - 9 | 1889 | 83 | |
| XM_015099149.2 | XP_014954635.1 | 8 - 9 | 1612 | 83 | ||
More Information
Set Membership: |
|

RO60
Genomic Map