Assay Details
| Gene Symbol: SYT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959600.1 | XP_027815401.1 | 5 - 6 | 1406 | 58 | |
| XM_027959601.1 | XP_027815402.1 | 6 - 7 | 1032 | 58 | ||
More Information
Set Membership: |
|

SYT12
Genomic Map