Assay Details
| Gene Symbol: LOC101116707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964869.1 | XP_027820670.1 | 2 - 3 | 183 | 70 | |
| XM_027964870.1 | XP_027820671.1 | 2 - 3 | 215 | 70 | ||
| XM_027964871.1 | XP_027820672.1 | 2 - 3 | 183 | 70 | ||
More Information
Set Membership: |
|

LOC101116707
Genomic Map