Assay Details
| Gene Symbol: LOC101121595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968820.1 | XP_027824621.1 | 9 - 10 | 2042 | 76 | |
| XM_027968821.1 | XP_027824622.1 | 9 - 10 | 2052 | 76 | ||
| XM_027968822.1 | XP_027824623.1 | 8 - 9 | 1957 | 76 | ||
| XM_027968823.1 | XP_027824624.1 | 9 - 10 | 1115 | 76 | ||
More Information
Set Membership: |
|

LOC101121595
Genomic Map