Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008777.4 | XP_004008826.2 | 11 - 12 | 1539 | 71 | |
| XM_012178453.2 | XP_012033843.2 | 11 - 12 | 1561 | 71 | ||
| XM_012178454.3 | XP_012033844.2 | 11 - 12 | 1460 | 71 | ||
| XM_027970187.1 | XP_027825988.1 | 11 - 12 | 1460 | 71 | ||
More Information
Set Membership: |
|

PWWP3A
Genomic Map