Assay Details
| Gene Symbol: LOC114115563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971515.1 | XP_027827316.1 | 3 - 4 | 424 | 108 | |
| Gene Symbol: LOC101111971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963821.1 | XP_027819622.1 | 8 - 9 | 1221 | 108 | |
More Information
Set Membership: |
|

LOC114115563
Genomic Map