Assay Details
| Gene Symbol: PDCD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978209.1 | XP_027834010.1 | 3 - 4 | 347 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDCD2L| Gene Symbol: PDCD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978209.1 | XP_027834010.1 | 3 - 4 | 347 | 61 | |
Set Membership: |
|