Assay Details
Gene Symbol: ZNF404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190498.3 | XP_012045888.1 | 5 - 6 | 555 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
