Assay Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970178.1 | XP_027825979.1 | 8 - 9 | 1679 | 58 | |
| XM_027970179.1 | XP_027825980.1 | 8 - 9 | 1680 | 58 | ||
| XM_027970180.1 | XP_027825981.1 | 7 - 8 | 1546 | 58 | ||
More Information
Set Membership: |
|

MIDN
Genomic Map