Assay Details
| Gene Symbol: C9H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973442.1 | XP_027829243.1 | 2 - 3 | 261 | 94 | |
| XM_027973443.1 | XP_027829244.1 | 3 - 4 | 848 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H8orf33| Gene Symbol: C9H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973442.1 | XP_027829243.1 | 2 - 3 | 261 | 94 | |
| XM_027973443.1 | XP_027829244.1 | 3 - 4 | 848 | 94 | ||