Assay Details
| Gene Symbol: LOC101117683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975249.1 | XP_027831050.1 | 8 - 9 | 1339 | 64 | |
| XM_027975250.1 | XP_027831051.1 | 8 - 9 | 2035 | 64 | ||
| XM_027975251.1 | XP_027831052.1 | 8 - 9 | 1338 | 64 | ||
| XM_027975252.1 | XP_027831053.1 | 8 - 9 | 1338 | 64 | ||
| XM_027975253.1 | XP_027831054.1 | 8 - 9 | 1338 | 64 | ||
| XM_027975254.1 | XP_027831055.1 | 8 - 9 | 1339 | 64 | ||
| XM_027975255.1 | XP_027831056.1 | 8 - 9 | 1339 | 64 | ||
| XM_027975256.1 | XP_027831057.1 | 7 - 8 | 1925 | 64 | ||
| XM_027975257.1 | XP_027831058.1 | 7 - 8 | 1924 | 64 | ||
More Information
Set Membership: |
|

LOC101117683
Genomic Map