Assay Details
| Gene Symbol: LOC114112551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963825.1 | XP_027819626.1 | 2 - 3 | 455 | 79 | |
| Gene Symbol: HSPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965547.1 | XP_027821348.1 | 4 - 5 | 424 | 79 | |

LOC114112551
Genomic Map