Assay Details
Gene Symbol: LOC101117600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013284.4 | XP_004013333.1 | 4 - 5 | 580 | 81 | |
XM_027975009.1 | XP_027830810.1 | 3 - 4 | 466 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
