Assay Details
| Gene Symbol: LOC101112469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967285.1 | XP_027823086.1 | 4 - 5 | 3419 | 63 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101112469| Gene Symbol: LOC101112469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967285.1 | XP_027823086.1 | 4 - 5 | 3419 | 63 | |