Assay Details
| Gene Symbol: PACSIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012175736.3 | XP_012031126.1 | 6 - 7 | 1457 | 58 | |
| XM_015095054.2 | XP_014950540.1 | 6 - 7 | 958 | 58 | ||
| XM_027968177.1 | XP_027823978.1 | 6 - 7 | 960 | 58 | ||
More Information
Set Membership: |
|

PACSIN2
Genomic Map