Assay Details
| Gene Symbol: IMPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011864.4 | XP_004011913.3 | 5 - 6 | 2162 | 75 | |
| XM_015097949.2 | XP_014953435.2 | 5 - 6 | 426 | 75 | ||
More Information
Set Membership: |
|

IMPA1
Genomic Map