Assay Details
| Gene Symbol: LOC101115527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006175.4 | XP_004006224.1 | 3 - 4 | 474 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115527| Gene Symbol: LOC101115527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006175.4 | XP_004006224.1 | 3 - 4 | 474 | 96 | |
Set Membership: |
|