Assay Details
| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 3 - 4 | 394 | 94 | |
| GenBank mRNA | FJ958364.1 | - | 3 - 4 | 193 | 94 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122608| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 3 - 4 | 394 | 94 | |
| GenBank mRNA | FJ958364.1 | - | 3 - 4 | 193 | 94 | |