Assay Details
| Gene Symbol: LOC101102259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015893.4 | XP_004015942.3 | 5 - 6 | 2798 | 66 | |
| GenBank mRNA | MF197687.1 | - | 2 - 3 | 350 | 66 | |
| MF197688.1 | - | 3 - 4 | 455 | 66 | ||
More Information
Set Membership: |
|

LOC101102259
Genomic Map