Assay Details
| Gene Symbol: LOC101102259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015893.4 | XP_004015942.3 | 6 - 7 | 2966 | 104 | |
| GenBank mRNA | MF197687.1 | - | 3 - 4 | 518 | 104 | |
| MF197688.1 | - | 4 - 5 | 623 | 104 | ||
More Information
Set Membership: |
|

LOC101102259
Genomic Map