Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968613.1 | XP_027824414.1 | 3 - 4 | 268 | 79 | |
| XM_027968614.1 | XP_027824415.1 | 3 - 4 | 259 | 79 | ||
More Information
Set Membership: |
|

IMMP2L
Genomic Map