Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097815.2 | XP_014953301.2 | 5 - 6 | 1115 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RDH10| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097815.2 | XP_014953301.2 | 5 - 6 | 1115 | 66 | |
Set Membership: |
|