Assay Details
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961166.1 | XP_027816967.1 | 5 - 6 | 2620 | 83 | |
| XM_027961168.1 | XP_027816969.1 | 6 - 7 | 1795 | 83 | ||
| XM_027961169.1 | XP_027816970.1 | 6 - 7 | 1810 | 83 | ||
More Information
Set Membership: |
|

PTCD1
Genomic Map