Assay Details
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961166.1 | XP_027816967.1 | 7 - 8 | 2918 | 76 | |
| XM_027961168.1 | XP_027816969.1 | 8 - 9 | 2093 | 76 | ||
| XM_027961169.1 | XP_027816970.1 | 8 - 9 | 2108 | 76 | ||
More Information
Set Membership: |
|

PTCD1
Genomic Map