Assay Details
| Gene Symbol: SMPDL3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005072.4 | XP_004005121.3 | 6 - 7 | 1383 | 67 | |
| XM_012150886.3 | XP_012006276.2 | 5 - 6 | 1264 | 67 | ||
More Information
Set Membership: |
|

SMPDL3B
Genomic Map