Assay Details
| Gene Symbol: RETREG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012940.4 | XP_004012989.1 | 8 - 9 | 1064 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RETREG3| Gene Symbol: RETREG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012940.4 | XP_004012989.1 | 8 - 9 | 1064 | 87 | |