Assay Details
| Gene Symbol: TRUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960242.1 | XP_027816043.1 | 7 - 8 | 874 | 84 | |
| XM_027960243.1 | XP_027816044.1 | 7 - 8 | 793 | 84 | ||
More Information
Set Membership: |
|

TRUB1
Genomic Map