Assay Details
| Gene Symbol: LOC114114046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967819.1 | XP_027823620.1 | 5 - 6 | 268 | 80 | |
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967817.1 | XP_027823618.1 | 16 - 17 | 1272 | 80 | |
More Information
Set Membership: |
|

LOC114114046
Genomic Map