Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967817.1 | XP_027823618.1 | 4 - 5 | 271 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRAG2| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967817.1 | XP_027823618.1 | 4 - 5 | 271 | 77 | |
Set Membership: |
|