Assay Details
Gene Symbol: C5H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177809.3 | XP_012033199.2 | 4 - 5 | 396 | 76 | |
XM_027969515.1 | XP_027825316.1 | 4 - 5 | 649 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
