Assay Details
| Gene Symbol: LOC101112261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980059.1 | XP_027835860.1 | 7 - 8 | 928 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101112261| Gene Symbol: LOC101112261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980059.1 | XP_027835860.1 | 7 - 8 | 928 | 81 | |
Set Membership: |
|