Assay Details
Gene Symbol: C21H11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959690.1 | XP_027815491.1 | 2 - 3 | 683 | 117 |

More Information
Set Membership: |
![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C21H11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959690.1 | XP_027815491.1 | 2 - 3 | 683 | 117 |
Set Membership: |
![]() ![]() |