Assay Details
| Gene Symbol: MYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004891.4 | XP_004004940.1 | 4 - 5 | 825 | 68 | |
| XM_004004892.4 | XP_004004941.1 | 4 - 5 | 487 | 68 | ||
| GenBank mRNA | KJ700419.1 | - | 4 - 5 | 410 | 68 | |
| KJ710701.1 | - | 4 - 5 | 607 | 68 | ||
More Information
Set Membership: |
|

MYL1
Genomic Map