Assay Details
| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 14 - 15 | 2926 | 93 | |
| XM_012182449.3 | XP_012037839.2 | 14 - 15 | 2929 | 93 | ||
| XM_012182451.3 | XP_012037841.2 | 14 - 15 | 2930 | 93 | ||

LOC101114198
Genomic Map