Assay Details
| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 9 - 10 | 1804 | 91 | |
| XM_012182449.3 | XP_012037839.2 | 9 - 10 | 1804 | 91 | ||
| XM_012182451.3 | XP_012037841.2 | 9 - 10 | 1805 | 91 | ||

LOC101114198
Genomic Map