Assay Details
| Gene Symbol: LOC101113054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965887.1 | XP_027821688.1 | 49 - 50 | 3670 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113054| Gene Symbol: LOC101113054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965887.1 | XP_027821688.1 | 49 - 50 | 3670 | 63 | |
Set Membership: |
|