Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013591.3 | XP_004013640.1 | 7 - 8 | 1439 | 68 | |
| XM_004013592.4 | XP_004013641.1 | 7 - 8 | 2217 | 68 | ||
| XM_012187013.2 | XP_012042403.1 | 7 - 8 | 2595 | 68 | ||
More Information
Set Membership: |
|

BRINP3
Genomic Map