Assay Details
| Gene Symbol: SMARCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017663.4 | XP_004017712.3 | 7 - 8 | 1133 | 74 | |
| XM_015101733.2 | XP_014957219.2 | 7 - 8 | 1166 | 74 | ||
More Information
Set Membership: |
|

SMARCB1
Genomic Map