Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018824.4 | XP_004018873.2 | 6 - 7 | 6319 | 116 | |
| XM_012100682.2 | XP_011956072.2 | 6 - 7 | 1986 | 116 | ||
| XM_015102717.2 | XP_014958203.2 | 6 - 7 | 2026 | 116 | ||
| XM_027958601.1 | XP_027814402.1 | 3 - 4 | 3865 | 116 | ||
More Information
Set Membership: |
|

BICRAL
Genomic Map