Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104655.2 | XP_014960141.1 | 2 - 3 | 770 | 78 | |
| XM_015104656.2 | XP_014960142.1 | 2 - 3 | 167 | 78 | ||
More Information
Set Membership: |
|

NKIRAS1
Genomic Map