Assay Details
| Gene Symbol: FRMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959397.1 | XP_027815198.1 | 10 - 11 | 1407 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRMD8| Gene Symbol: FRMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959397.1 | XP_027815198.1 | 10 - 11 | 1407 | 58 | |
Set Membership: |
|